Order Entry
Switzerland
ContactUsLinkComponent
 
Toxic
Health
4-Nitroaniline ≥99.0% (by HPLC) indicator in non-aqueous solvents, Fluka™
Catalog # 72680-500G
CAS Number:  
Toxic
Health
4-Nitroaniline ≥99.0% (by HPLC) indicator in non-aqueous solvents, Fluka™
Catalog # 72680-500G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (HPLC):
    Min. 99.0 %
  • Melting point:
    146 - 150 °C
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • Residue on ignition:
    Max. 0.05 %

Specification Test Results

About this item

 :
 :138.13 g/mol
Merck Index:12,06681
Boiling Pt:260 °C
Melting Pt:146-149 °C
Melting Pt:146…149 °C
Density:1.44 g/cm³
Flash Pt:213 °C
Storage Temperature:Ambient
EINECS:202-810-1
UN:1661
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:100-01-6
Formula:C₆H₆N₂O₂
MDL Number:MFCD00007858