

N,N-Dimethyl-4-nitrosoaniline for synthesis, Sigma-Aldrich®
Catalog # 8.20515.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:84 - 87 °C
- Molar Mass:150.18 g/mol
- Density:1.15 g/cm3 (20 °C)
Specification Test Results
About this item
:
:150.18 g/mol
Merck Index:12,06736
Boiling Pt:259 °C (1013 hPa)
Melting Pt:87 °C
Density:1.145 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-343-1
UN:1369
ADR:4.2,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS06
CAS Number:138-89-6
Formula:C₈H₁₀N₂O
MDL Number:MFCD00002063