Orders LinkContactUsLinkComponent
Filter discs, DURAN®
Supplier:   DWK Life Sciences
Filter discs, DURAN®
Supplier:  DWK Life Sciences
Certificates
Filter discs, DURAN®
Filter discs, DURAN®
62 Options Available Below
Product Details & Documents
Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.
  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design
Made from clear DURAN® borosilicate glass 3,3.
Product Family Options
Product Information
  • Porosity
    Ø
    Thickness
    Availability
    Price
  • 0
    10 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 64.60
    SFr. 64.60
    1 * 10 ST
       SFr. 64.60
  • 0
    20 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 67.10
    SFr. 67.10
    1 * 10 ST
       SFr. 67.10
  • 0
    25 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 68.60
    SFr. 68.60
    1 * 10 ST
       SFr. 68.60
  • 0
    30 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 75.80
    SFr. 75.80
    1 * 10 ST
       SFr. 75.80
  • 0
    40 mm
    4,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 98.60
    SFr. 98.60
    1 * 10 ST
       SFr. 98.60
  • 0
    50 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 65.40
    SFr. 65.40
    1 * 5 ST
       SFr. 65.40
  • 0
    55 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 19.40
    SFr. 19.40
    1 * 1 ST
       SFr. 19.40
  • 0
    60 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 21.00
    SFr. 21.00
    1 * 1 ST
       SFr. 21.00
  • 0
    70 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 36.70
    SFr. 36.70
    1 * 1 ST
       SFr. 36.70
  • 0
    90 mm
    7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 45.40
    SFr. 45.40
    1 * 1 ST
       SFr. 45.40
  • 0
    120 mm
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 109.00
    SFr. 109.00
    1 * 1 ST
       SFr. 109.00
  • 1
    10 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 63.70
    SFr. 63.70
    1 * 10 ST
       SFr. 63.70
  • 1
    20 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 66.20
    SFr. 66.20
    1 * 10 ST
       SFr. 66.20
  • 1
    25 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 67.70
    SFr. 67.70
    1 * 10 ST
       SFr. 67.70
  • 1
    30 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 74.60
    SFr. 74.60
    1 * 10 ST
       SFr. 74.60
  • 1
    40 mm
    3,8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 97.10
    SFr. 97.10
    1 * 10 ST
       SFr. 97.10
  • 1
    50 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 65.40
    SFr. 65.40
    1 * 5 ST
       SFr. 65.40
  • 1
    55 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 19.10
    SFr. 19.10
    1 * 1 ST
       SFr. 19.10
  • 1
    60 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 20.70
    SFr. 20.70
    1 * 1 ST
       SFr. 20.70
  • 1
    70 mm
    5,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 36.10
    SFr. 36.10
    1 * 1 ST
       SFr. 36.10
  • 1
    90 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 44.70
    SFr. 44.70
    1 * 1 ST
       SFr. 44.70
  • 1
    120 mm
    8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 107.00
    SFr. 107.00
    1 * 1 ST
       SFr. 107.00
  • 2
    10 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 63.70
    SFr. 63.70
    1 * 10 ST
       SFr. 63.70
  • 2
    15 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 65.60
    SFr. 65.60
    1 * 10 ST
       SFr. 65.60
  • 2
    20 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 66.20
    SFr. 66.20
    1 * 10 ST
       SFr. 66.20
  • 2
    25 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 67.70
    SFr. 67.70
    1 * 10 ST
       SFr. 67.70
  • 2
    30 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 74.60
    SFr. 74.60
    1 * 10 ST
       SFr. 74.60
  • 2
    40 mm
    3,8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 97.10
    SFr. 97.10
    1 * 10 ST
       SFr. 97.10
  • 2
    50 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 65.40
    SFr. 65.40
    1 * 5 ST
       SFr. 65.40
  • 2
    55 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 19.10
    SFr. 19.10
    1 * 1 ST
       SFr. 19.10
  • 2
    60 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 20.70
    SFr. 20.70
    1 * 1 ST
       SFr. 20.70
  • 2
    70 mm
    5,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 36.10
    SFr. 36.10
    1 * 1 ST
       SFr. 36.10
  • 2
    90 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 44.70
    SFr. 44.70
    1 * 1 ST
       SFr. 44.70
  • 2
    120 mm
    8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 107.00
    SFr. 107.00
    1 * 1 ST
       SFr. 107.00
  • 3
    10 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 63.70
    SFr. 63.70
    1 * 10 ST
       SFr. 63.70
  • 3
    20 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 66.20
    SFr. 66.20
    1 * 10 ST
       SFr. 66.20
  • 3
    25 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 67.70
    SFr. 67.70
    1 * 10 ST
       SFr. 67.70
  • 3
    30 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 74.60
    SFr. 74.60
    1 * 10 ST
       SFr. 74.60
  • 3
    40 mm
    3,8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 97.10
    SFr. 97.10
    1 * 10 ST
       SFr. 97.10
  • 3
    50 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 65.40
    SFr. 65.40
    1 * 5 ST
       SFr. 65.40
  • 3
    55 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 19.10
    SFr. 19.10
    1 * 1 ST
       SFr. 19.10
  • 3
    60 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 20.70
    SFr. 20.70
    1 * 1 ST
       SFr. 20.70
  • 3
    70 mm
    5,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 36.10
    SFr. 36.10
    1 * 1 ST
       SFr. 36.10
  • 3
    90 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 44.70
    SFr. 44.70
    1 * 1 ST
       SFr. 44.70
  • 3
    120 mm
    8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 107.00
    SFr. 107.00
    1 * 1 ST
       SFr. 107.00
  • 4
    10 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 72.20
    SFr. 72.20
    1 * 10 ST
       SFr. 72.20
  • 4
    15 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 7.50
    SFr. 7.50
    1 * 1 ST
       SFr. 7.50
  • 4
    20 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 74.60
    SFr. 74.60
    1 * 10 ST
       SFr. 74.60
  • 4
    25 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 78.30
    SFr. 78.30
    1 * 10 ST
       SFr. 78.30
  • 4
    30 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 88.20
    SFr. 88.20
    1 * 10 ST
       SFr. 88.20
  • 4
    40 mm
    3,8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 117.00
    SFr. 117.00
    1 * 10 ST
       SFr. 117.00
  • 4
    50 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 75.90
    SFr. 75.90
    1 * 5 ST
       SFr. 75.90
  • 4
    55 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 23.20
    SFr. 23.20
    1 * 1 ST
       SFr. 23.20
  • 4
    60 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 25.90
    SFr. 25.90
    1 * 1 ST
       SFr. 25.90
  • 4
    70 mm
    5,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 46.60
    SFr. 46.60
    1 * 1 ST
       SFr. 46.60
  • 4
    90 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 58.70
    SFr. 58.70
    1 * 1 ST
       SFr. 58.70
  • 4
    120 mm
    8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 142.00
    SFr. 142.00
    1 * 1 ST
       SFr. 142.00
  • 5
    15 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 216.00
    SFr. 216.00
    1 * 10 ST
       SFr. 216.00
  • 5
    25 mm
    3,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 294.00
    SFr. 294.00
    1 * 10 ST
       SFr. 294.00
  • 5
    40 mm
    3,8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 766.00
    SFr. 766.00
    1 * 10 ST
       SFr. 766.00
  • 5
    50 mm
    4,2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 237.00
    SFr. 237.00
    1 * 5 ST
       SFr. 237.00
  • 5
    120 mm
    8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 293.00
    SFr. 293.00
    1 * 1 ST
       SFr. 293.00