Order Entry
Switzerland
Orders LinkContactUsLinkComponent
3-Fluoroaniline

3-Fluoroaniline

Glass bottle for solids, 100 g
Catalog # MOLE35787562-100G
Irritant
Marketsource
Supplier:   Molekula
3-Fluoroaniline
Catalog # MOLE35787562-100G
Supplier:  Molekula
Marketsource
CAS Number:  372-19-0
3-Fluoroaniline

Glass bottle for solids, 100 g

CAS Number:  372-19-0
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:372-19-0
  • Formula:C₆H₆FN
  • MDL Number:MFCD00007758
  • Molecular Weight:111.12 g/mol
  • Boiling Pt:186 °C (1013 hPa)
  • Melting Pt:–2 °C
  • Density:1.16 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-747-0
  • UN:2941
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.