
3-Fluoro-p-anisaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:351-54-2
:
Formula:C₈H₇FO₂
MDL Number:MFCD00003349
:154.14 g/mol
Boiling Pt:129…132 °C (11 mmHg)
Melting Pt:34…35 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:206-514-3
:GHS07
Related Information
Frequently Bought Together