Orders LinkContactUsLinkComponent
Magnetic stirring bars, triangular
Magnetic stirring bars, triangular
Supplier:   KARTELL
Magnetic stirring bars, triangular
Supplier:  KARTELL
Certificates
Magnetic stirring bars, triangular
Magnetic stirring bars, triangular
4 Options Available Below
Product Details & Documents
PTFE coated, triangular magnetic stirring bars.
  • Scraping action ideal for dissolving solids and mixing sediments
  • Angled surfaces create high turbulence even at low speeds
  • Suitable for food stuff
Product Family Options
Product Information
  • Length
    Ø
    Availability
    Price
  • 12,21 mm
    5,61 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6.00
    SFr. 6.00
    1 * 1 ST
       SFr. 6.00
  • 25,32
    7,39 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 7.70
    SFr. 7.70
    1 * 1 ST
       SFr. 7.70
  • 33,83
    8,13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.90
    SFr. 9.90
    1 * 1 ST
       SFr. 9.90
  • 49,61
    10,43 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 13.40
    SFr. 13.40
    1 * 1 ST
       SFr. 13.40
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..