Bottles, wide neck, square, with screw cap
Supplier: BURKLE
Certificates
About this item
HDPE, transparent.
- Tamper evident blue screw cap
- Cap absolutely tight closing
- UN approval for bulk goods (except bottle with 50 ml capacity)
- Temperature resistance from −50 °C up to +110 C°
- Suitable for use with foodstuffs
The reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.
Frequently Bought Together