ContactUsLinkComponent
Adapters, receiver, bent, DURAN®
Adapters, receiver, bent, DURAN®
Catalog # SCOT243100808
Supplier:  DWK Life Sciences
undefined
Adapters, receiver, bent, DURAN®
Catalog # SCOT243100808
Supplier:  DWK Life Sciences
Specifications
  • Socket size:
    24/29 NS
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bent, with ground socket.