3-Chloro-4-fluorobenzoic acid
Catalog # MOLEM74233060
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:174.56 g/mol
CAS Number:403-16-7
Formula:C₇H₄ClFO₂
MDL Number:MFCD00042477
Boiling Pt:273
Melting Pt:136