
2-Chloro-6-fluorobenzoic acid
Catalog # MOLEM82871027
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:174.56 g/mol
Boiling Pt:272 °C (1013 hPa)
Melting Pt:157…159 °C
EINECS:207-105-2
:GHS07
Storage Temperature:Ambient
CAS Number:434-75-3
Formula:C₇H₄ClFO₂
MDL Number:MFCD00002417