ContactUsLinkComponent
Cleanroom wipes, AlphaLite® II
Cleanroom wipes, AlphaLite® II
Catalog # TEXWTX1091
Supplier:  Texwipe
undefined
Cleanroom wipes, AlphaLite® II
Catalog # TEXWTX1091
Supplier:  Texwipe
Specifications
  • Packed:
    150 wipes/bag, 4 bags/case
  • Description:
    TX1091
  • L×W:
    250×580 mm
  • Seq:
    02
  • Pk:
    600
Specifications
About this item
These wipes are primarily intended for the usage as flat mop covers for cleaning and decontaminating floors. Made of non-woven polyester, they are especially suitable for cleaning and removing residues.
  • Knit polyester for applying disinfectants and biocides
  • High absorbency and excellent abrasion resistance
Suitable for use in ISO cleanroom class 3 - 8 environments.