Order Entry
Switzerland
ContactUsLinkComponent
 
Chlorobenzene, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
 
undefined
Chlorobenzene, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
 

 

  • Assay (by GC):
    min. 99 %
  • Boiling Range:
    max. 1.0 °C
  • Density (g/ml) at 25°C:
    1.101 - 1.104
  • Recorded Boiling Point:
    132 °C

 

 

Frequently Bought Together