L-shaped spreaders, PS
Supplier: KARTELL
Certificates
About this item
Disposable L-shaped sterile PS spreaders for bacteriology with special 100° angling for easier use. Especially handy for dispersing liquids across agar culture plates.
Packaging: Available single-wrapped or in packs of 5.
Frequently Bought Together