Orders LinkContactUsLinkComponent
Burettes, Pellet pattern, without intermediate stopcock, Schellbach
Supplier:   HIRSCHMANN
Burettes, Pellet pattern, without intermediate stopcock, Schellbach
Supplier:  HIRSCHMANN
Certificates
Burettes, Pellet pattern, without intermediate stopcock, Schellbach
Burettes, Pellet pattern, without intermediate stopcock, Schellbach
6 Options Available Below
Product Details & Documents
Automatic burette, Pellet, class AS, DURAN® with blue graduation.
  • Lateral stopcock with glass key or PTFE spindle
  • Schellbach stripes for enhanced legibility
  • Certified conformity, with dated batch identification
 : Compliance with DIN EN ISO 385.
Product Family Options
Lateral stopcock, with glass key, without reservoir bottle
Product Information
  • Capacity
    Division
    Scale length
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    500 mm
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 299.00
    SFr. 299.00
    1 * 1 ST
       SFr. 299.00
  • 25 ml
    0,05 ml
    500 mm
    0,045 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 299.00
    SFr. 299.00
    1 * 1 ST
       SFr. 299.00
  • 50 ml
    0,1 ml
    500 mm
    0,075 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 299.00
    SFr. 299.00
    1 * 1 ST
       SFr. 299.00
Lateral stopcock, with PTFE spindle, without reservoir bottle
Product Information
  • Capacity
    Division
    Scale length
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    500 mm
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 337.00
    SFr. 337.00
    1 * 1 ST
       SFr. 337.00
  • 25 ml
    0,05 ml
    500 mm
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 337.00
    SFr. 337.00
    1 * 1 ST
       SFr. 337.00
  • 50 ml
    0,1 ml
    500 mm
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 337.00
    SFr. 337.00
    1 * 1 ST
       SFr. 337.00
Reservoir bottle
Product Information
  • Catalog #
    Description
    Capacity
    Availability
    Price
  • Reservoir bottle, NS 29/32, DURAN® clear glass
    2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 36.70
    SFr. 36.70
    1 * 1 ST
       SFr. 36.70