GC septa, long life GC autosampler septa
Long Life GC autosampler septa are designed for use with GC autosamplers.
- Inlet temperature of up to 400 °C
- Injection life of 400+ injections can be achieved with proper care
The 5, 9, 11 and 11,5 mm diameter septa are available with a unique prepierced center depression which guides the syringe needle to the same injection point. The 9,5, 12,7 and 17 mm diameter septa are supplied without the unique center depression.
Frequently Bought Together