Storm water samplers, Nalgene®
About this item
Storm water samplers with HDPE bottle.
- Leakproof when closure is applied
- Device intended for one-time use
Suitable for collecting samples for inorganic analyses.
Certifications: EPA NPDES MSGP.
Caution: Not for oil and grease sample collection.
*The product(s) on this page are outside the scope of the EU Directive 2019/904
3 Options Available Below
Product Details & Documents
Storm water samplers with HDPE bottle.
- Leakproof when closure is applied
- Device intended for one-time use
Suitable for collecting samples for inorganic analyses.
Certifications: EPA NPDES MSGP.
Caution: Not for oil and grease sample collection.
*The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications for Product Family
Specifications
- Catalog No:
- NALG1100-1000
- NALG1120-1000
- NALG1160-1000
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
- CapacityAvailabilityPrice
- NALG1100-1000Storm water samplersSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.NALG1100-1000Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - NALG1120-1000Storm water samplerSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.NALG1120-1000Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - NALG1160-1000Storm water mounting kitSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.NALG1160-1000Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact