BD Nokor™ Admix Needles
Anbieter: BD Medical
Zertifikate
Über diesen Artikel
The BD Nokor™ Admix Needles are single-use medical devices used for the aspiration of fluids from vials and ampoules. These needles are not intended for skin injections.
- Vented point design features a cannula side port
- Design allows fluid placement on the container sidewall during reconstitution, helping reduce foaming or spray
- Sterile, single-use design
The BD Nokor™ Admix needles has a cannula side port to place fluid on the sidewall of a container during reconstitution that helps reduce foaming or spray of powdered medication.
Frequently Bought Together