Order Entry
Switzerland
ContactUsLinkComponent
N-Methylanthranilic acid for synthesis, Sigma-Aldrich®
N-Methylanthranilic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41325.0100
Supplier:  Merck
CAS Number:  
N-Methylanthranilic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41325.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    0.20 g/l (20 °C)
  • Melting Point:
    170 - 175 °C
  • Molar Mass:
    151.16 g/mol

Specification Test Results

About this item

 :
 :151.16 g/mol
Boiling Pt:80 °C (0.01 mmHg)
Melting Pt:172 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-343-9
 :GHS07
CAS Number:119-68-6
Formula:C₈H₉NO₂
MDL Number:MFCD00002424