Order Entry
Switzerland
ContactUsLinkComponent
Benzoic acid ≥99.7%, AnalaR NORMAPUR® analytical reagent
Health
Corrosive
Benzoic acid ≥99.7%, AnalaR NORMAPUR® analytical reagent
Catalog # 20172.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Corrosive
Benzoic acid ≥99.7%, AnalaR NORMAPUR® analytical reagent
Catalog # 20172.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99.7 %
  • Substances reducing KMnO4 (as O):
    Passes test
  • Substances coloured by H2SO4:
    Passes test
  • Melting point:
    121 - 123 °C
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in absolute ethanol:
    Max. 50 ppm
  • Insolubility in diluted ammonia:
    Max. 50 ppm
  • Insolubility in NaOH 1 N:
    Max. 50 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Total Cl (Chlorine):
    Max. 50 ppm
  • Water:
    Max. 0.3 %
  • SO4 (Sulphate):
    Max. 30 ppm
  • Fe (Iron):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
 :GHS08
 :GHS05
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398