

Benzoic acid, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
: Merck
:65-85-0
:
:C₇H₆O₂
:MFCD00002398
:122.12 g/mol
:13,01092
:249 °C (1013 hPa)
:121…123 °C
:1.316 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:121 °C (closed cup)
:Ambient
:200-618-2
:01-2119455536-33
:GHS05
:GHS08
- Solubility:2.9 g/l (25 °C)
- Melting Point:121 - 123 °C
- Molar Mass:122.12 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:249 °C (1013 hPa)
- Vapor Pressure:0.001 hPa (20 °C)
- Flash Point:121 °C
- Density:1.321 g/cm3 (20 °C)
- pH:2.5 - 3.5 (H2O, 20 °C) (saturated solution)
- Ignition Point:570 °C
Frequently Bought Together