

Benzoic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65-85-0
:
Formula:C₇H₆O₂
MDL Number:MFCD00002398
:122.12 g/mol
Merck Index:14,1091
Boiling Pt:249 °C
Melting Pt:121…124 °C
Density:1.32
Flash Pt:121 °C (250 °F)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
:GHS05
:GHS07
Related Information
Frequently Bought Together