Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ethyl 4-hydroxybenzoate

Ethyl 4-hydroxybenzoate

Glass bottle for solids, 150 mg
Catalog # EDQME2425000
Supplier:   EDQM
Ethyl 4-hydroxybenzoate
Catalog # EDQME2425000
Supplier:  EDQM
CAS Number:  120-47-8
Ethyl 4-hydroxybenzoate

Glass bottle for solids, 150 mg

CAS Number:  120-47-8
Price
SFr. 245.00
Log in for availability
1 * 150 mg
Product Details & Documents
Synonyms:  Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
  • CAS Number:120-47-8
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002353
  • Molecular Weight:166.18 g/mol
  • Merck Index:13,03869
  • Boiling Pt:297 °C (1013 hPa)
  • Melting Pt:116…118 °C
  • Density:1.078 g/cm³ (131 °C)
  • Storage Temperature:Ambient
  • EINECS:204-399-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    150 mg

Specifications