Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ethyl 4-hydroxybenzoate ≥99%

Ethyl 4-hydroxybenzoate ≥99%

Ethyl 4-hydroxybenzoate ≥99%
Certificates
Ethyl 4-hydroxybenzoate  ≥99%
Ethyl 4-hydroxybenzoate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 120-47-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 116-118° Boiling Point: 297-298° dec. Merck Reference: 14,3837 MDL No.: MFCD00002353

Synonyms:  Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
  • CAS Number:120-47-8
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002353
  • Molecular Weight:166.18 g/mol
  • Merck Index:14,3837
  • Boiling Pt:297…298 °C (dec.)
  • Melting Pt:116…118 °C
  • Density:1.078 g/cm³ (131 °C)
  • Storage Temperature:Ambient
  • EINECS:204-399-4
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.