Orders LinkContactUsLinkComponent
Worktables, motorised, Concept
Supplier:   TRESTON
Worktables, motorised, Concept
Supplier:  TRESTON
Certificates
Worktables, motorised, Concept
39 Options Available Below
Product Details & Documents
The motorised sit-stand worktable is designed to meet the ergonomic requirements of today's labs; it adapts to the work process and the needs of the user.
  • The working position can be changed from 700 to 1100 mm
  • Loading capacity is max. 400 kg
  • Control panel includes up, down
  • 3 memory buttons for storing pre-selected heights
  • Frame, tabletop, and accessories available with ESD protection
  • Colour - Light grey
The sit-stand worktable can be used as a flat workspace or in combination with heavy equipment such as laminar flow cabinets. Even suitable for clean room ISO class 7.
 : Bench top needs to be ordered separately. A wide range of ergonomic accessories is available, such as adjustable shelves, LCD arms and keyboard holders. For details, please contact your local Avantor sales office.
Product Family Options
ESD worktable frames, motor adjustable (frame only)
Product Information
  • Material
    W×D×H
    Availability
    Price
  • SOVE10349015P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1000×600×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349016P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1200×600×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349017P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1500×600×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3’140.00
    SFr. 3’140.00
    1 * 1 ST
       SFr. 3’140.00
  • SOVE10349018P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1800×600×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349030P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1000×750×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349032P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1500×750×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349033P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1800×750×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349062P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1000×900×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349063P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1200×900×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349064P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1500×900×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349065P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    1800×900×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVE10349066P
    Concept worktable frame, height adjustable with motor
    Concept worktable frame, height adjustable with motor
    Epoxy powder coated steel
    2000×900×(700 - 1100) mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Concept tabletop, High pressure laminate
Product Information
  • Material
    W×D×H
    Availability
    Price
  • SOVETT10060-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1000×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT10075-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1000×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT10090-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1000×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT12060-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1200×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT12090-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1200×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT15060-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1500×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT15075-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1500×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT18060-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1806×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT18075-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1806×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT18090-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    1806×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT20060-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    2000×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT20090-HPL
    Concept tabletop, High pressure laminate
    Concept tabletop, High pressure laminate
    Chipboard
    2000×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Concept tabletop, ESD
Product Information
  • Material
    W×D×H
    Availability
    Price
  • SOVETT10060-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1000×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT10075-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1000×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT10090-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1000×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT12060-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1200×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT12075-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1200×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT12090-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1200×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT15060-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1500×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT15075-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1500×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT18060-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1806×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT18075-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1806×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT18090-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    1806×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT20060-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    2000×600×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT20075-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    2000×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOVETT20090-ESD
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Tabletop for Concept table, ESD, Semi-conductive laminated
    Chipboard
    2000×900×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Concept tabletop, Low pressure laminate
Product Information
  • Material
    W×D×H
    Availability
    Price
  • SOVETT20075-LPL
    Concept tabletop, Low pressure laminate
    Concept tabletop, Low pressure laminate
    Chipboard
    2000×750×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 224.00
    SFr. 224.00
    1 * 1 ST
       SFr. 224.00