Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, p-Anisaldehyde
Supelco® Organic Reference Standard, p-Anisaldehyde
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, p-Anisaldehyde
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    123-11-5
  • Storage conditions:
    RT
  • Hazard class description:
    H412
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00003385
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4-Methoxybenzaldehyde; Aubépine
  • Melting point:
    –1 °C (lit.)
  • Formula:
    CH₃OC₆H₄CHO
  • Molecular weight:
    136,15
  • Density:
    1,119 g/ml at 25 °C (lit.)
  • Flash point:
    116 °C - Closed cup
  • Boiling point:
    248 °C (lit.)

Specifications

Related Information

Frequently Bought Together