ContactUsLinkComponent
Staining jars according to Hellendahl, DURAN®
Staining jars according to Hellendahl, DURAN®
Catalog # SCOT233140001
Supplier:  DWK Life Sciences
undefined
Staining jars according to Hellendahl, DURAN®
Catalog # SCOT233140001
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Staining dishes acc. to Hellendahl, for up to 16 microscope slides
  • W×D×H:
    90×40×90 mm
  • Pk:
    10
Specifications
About this item
Soda-Lime glass.
For up to 16 microscope slides (76x26 mm).
Caution: Note: Do not clean staining dishes and jars at temperatures above 60°C (glass corrosion is possible).