Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Kovac's indole reagent, Millipore®

Kovac's indole reagent, Millipore®

Glass bottle, 100 mL
Catalog # SIAL60983-100ML
Supplier:   Merck
Kovac's indole reagent, Millipore®
Catalog # SIAL60983-100ML
Supplier:  Merck
CAS Number:  100-10-7
Kovac's indole reagent, Millipore®
Kovac's indole reagent, Millipore®

Glass bottle, 100 mL

CAS Number:  100-10-7
Price
SFr. 120.00
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  4-(Dimethylamino)benzaldehyde solution
  • CAS Number:100-10-7
  • Formula:(CH₃)₂NC₆H₄CHO
  • MDL Number:MFCD00003381
  • Molecular Weight:149.19 g/mol
  • Merck Index:13,03257
  • Boiling Pt:176…177 °C (1013 hPa)
  • Melting Pt:72 °C
  • Density:1.0254 g/cm³ (99.9 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:202-819-0

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.