Cleanroom floor wipes, sterile, AlphaLite® II
115-1893
: Texwipe
- Packed:10 wipes/bag, 34 bags/case
- Description:TX3424
- L×W:600×250 mm
- Pk:340
Cut-edge polyester single ply wipe that is ideal for floor cleaning and for applying cleaning solutions and disinfectants on floors.
- Large size for improved application on floor surfaces
- Size is compatible with most mopping systems
- Improved contamination control
Suitable for use in Class 100, ISO 5 and Class 1000, ISO 6 environments.
: Packaging is adapted for sterile and highly critical environments.