Bell Jars, with knob, DURAN®
Nome del fornitore: DWK Life Sciences
Certificati
Informazioni su questo articolo
DURAN® boroslicate glass 3.3, clear.
- Good chemical and physical properties
- Excellent thermal resistance
- Quick and easy to clean
Wall thickness and geometry designed to suit vacuum applications.
Flat flange bell with knob and ground flange for mounting on a vacuum experimental plate (vacuum chamber).
Frequently Bought Together