Bottles, wide neck, HDPE, economy, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent with PP screw cap.
- Good chemical resistance
- Heat resistant from −100 to +120 °C
- Leakproof
Economical, break resistant disposable bottles for safe sample collection, transport and storage. Suitable for long-term storage in the absence of direct UV radiation.
Specifications
- Catalog No:
- 331-4261
- 331-4264
- 331-4263
- 331-4262
- 331-0060
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label