Bottles, narrow neck, round, HDPE, Boston type, with thread, Nalgene®
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
HDPE, translucent with PP screw cap.
- Good chemical resistance
- Heat resistant from −100 to +120 °C
- Leakproof
Ideal for storing, packaging and transporting liquids. Robust, strong and stable.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Spécifications
- Catalog No:
- NALG362002-0004
- NALG362002-0008
- NALG362002-0016
- NALG362002-0001
- NALG362002-0002
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Spécifications
Frequently Bought Together