Diagnostics bottles, narrow neck, round, without screw caps Nalgene®
Bottles without closure use in sample storage and the shipping of small volume, valuable reagents.
- Nonsterile
- Natural translucent PPCO provides excellent chemical resistance
- Durable and feature uniform walls resistant to splitting or puncturing
- Leakproof when assembled with closure
: Compliance with ISO 13485:2003 and 21CFR177.1520, USP Class VI.
: Coloured closures are ordered seperately.
- Catalog No:
- NALG362006-9125
- NALG362006-9025
- NALG362006-9050
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together