Order Entry
Switzerland
ContactUsLinkComponent
2,6-Dibromophenol
2,6-Dibromophenol
Catalog # EHERC12241200
Supplier:  EHRENSTORFER
CAS Number:  
2,6-Dibromophenol
Catalog # EHERC12241200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1,3-Dibromo-2-hydroxybenzene
 :
 :251.91 g/mol
Boiling Pt:255 °C (1013 hPa)
Melting Pt:55…56 °C
Storage Temperature:Ambient
EINECS:210-161-0
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:608-33-3
Formula:C₆H₄Br₂O
MDL Number:MFCD00002152