2-Butyl-1,3-propanediol
Catalog # WAKO323-31051
Supplier: Wako
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:132.2 g/mol
CAS Number:2612-26-2
Formula:C₇H₁₆O₂
MDL Number:MFCD00799395