Order Entry
Switzerland
ContactUsLinkComponent
Oxalic acid concentrate for dilution to 1L, 0.05 M (COOH)2 (0.1N) volumetric solution, Fluka™
Corrosive
Oxalic acid concentrate for dilution to 1L, 0.05 M (COOH)2 (0.1N) volumetric solution, Fluka™
Catalog # 38250-1EA
CAS Number:  
Corrosive
Oxalic acid concentrate for dilution to 1L, 0.05 M (COOH)2 (0.1N) volumetric solution, Fluka™
Catalog # 38250-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :90.04 g/mol
Boiling Pt:100 °C
Density:1.02 g/cm³
Storage Temperature:Ambient
ADR:8,III
 :Dose for preparation of 1 litre of solution.
EINECS:200-550-3
 :GHS05
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573