Order Entry
Switzerland
ContactUsLinkComponent
Sulphuric acid 100 g/L (∼9.6%)
Irritant
Sulphuric acid 100 g/L (∼9.6%)
Catalog # REGESU10WV1
Supplier:  REAGECON
CAS Number:  
Irritant
Sulphuric acid 100 g/L (∼9.6%)
Catalog # REGESU10WV1
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :98.08 g/mol
Merck Index:13,09064
Boiling Pt:101 °C (1013 hPa)
Density:1.066 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS07
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589