Organic reference standard, (±)-4-Vinylcyclohexene 2000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA17923100ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:100-40-3
- Description:(±)-4-Vinylcyclohexene 2,000 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₈H₁₂
- Description:(±)-4-Vinylcyclohexene