Organic reference standard, Fluorobenzene 2,000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA13781000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:462-06-6
- Description:Fluorobenzene 2,000 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₆H₅F
- Description:Fluorobenzene