Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 2,3,5-Trimethylphenol 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 2,3,5-Trimethylphenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17883500ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,3,5-Trimethylphenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17883500ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    697-82-5
  • Description:
    2,3,5-Trimethylphenol 100 µg/ml in methanol
  • Formula:
    C₉H₁₂O
  • Description:
    2,3,5-Trimethylphenol

Specifications

About this item