Pesticide reference standard, 2,4,6-Trichloroanisole 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA17714600ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:87-40-1
- Description:2,4,6-Trichloroanisole 100 µg/ml in methanol
- Formula:C₇H₅Cl₃O
- Description:2,4,6-Trichloroanisole