Organic reference standard, Methyl methacrylate (monomer) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA14971770CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:80-62-6
- Description:Methyl methacrylate (monomer) 100 µg/ml in cyclohexane
- Formula:C₅H₈O₂
- Description:Methyl methacrylate (monomer)