Organic reference standard, 2-Nitro-m-xylene 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA12728000ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:81-20-9
- Description:2-Nitro-m-xylene 100 µg/ml in methanol
- Formula:C₈H₉NO₂
- Description:2-Nitro-m-xylene