Organic reference standard, Chloromethane (Methyl chloride) 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA11419000ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74-87-3
- Description:Chloromethane (Methyl chloride) 100 µg/ml in methanol
- Formula:CH₃Cl
- Description:Chloromethane (methyl chloride)