Organic reference standard, chrysene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20670000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:218-01-9
- Description:Chrysene 10 µg/ml in cyclohexane
- Environmentally Preferable:
- Formula:C₁₈H₁₂
- Description:Chrysene