Pesticide reference standard, γ-BHC 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL14073000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:58-89-9
- Description:γ-BHC 10 µg/ml in cyclohexane
- Formula:C₆H₆Cl₆
- Description:γ-BHC