
Nitrosium hexafluorophosphate ≥96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16921-91-8
:
Formula:F₆NOP
MDL Number:MFCD00040324
:174.97 g/mol
Storage Temperature:Ambient
EINECS:240-980-9
UN:3260
ADR:8,II
:GHS05
Related Information
Frequently Bought Together