ContactUsLinkComponent
Snap cap for round bottom tubes, Falcon®
Snap cap for round bottom tubes, Falcon®
Catalog # 734-1303
Supplier:  Corning
   Low Manufacturing Impact
undefined
Snap cap for round bottom tubes, Falcon®
Catalog # 734-1303
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • For tubes:
    12×75 mm
  • Description:
    Snap cap, dual position, sterile, LDPE
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Pk:
    2.000
Specifications
About this item
Falcon® snap caps are designed for use with 12×75 mm test tubes
These polyethylene caps feature a patented, two-position fit, loose for aerobic culturing, tight for sealing. Sterile.
Ordering information: Snap caps are packed 500 per pack, 2000 per case.