Orders LinkContactUsLinkComponent
Screw caps for laboratory bottles
Screw caps for laboratory bottles
Supplier:   Lenz Laborglas
Screw caps for laboratory bottles
Supplier:  Lenz Laborglas
Certificates
Screw caps for laboratory bottles
Screw caps for laboratory bottles
3 Options Available Below
Product Details & Documents
PBT, red, with PTFE coated seals.
  • For DIN GL threads
  • Heat resistant up to +180 °C
  • Autoclavable, sterilisable with hot air
Product Family Options
Product Information
  • Thread
    Availability
    Price
  • GL 25
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 53.50
    SFr. 53.50
    1 * 10 ST
       SFr. 53.50
  • GL 32
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 80.80
    SFr. 80.80
    1 * 10 ST
       SFr. 80.80
  • GL 45
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 121.00
    SFr. 121.00
    1 * 10 ST
       SFr. 121.00