Burette stopcocks, lateral or straight
Supplier: Brand
Certificates
About this item
Borosilicate glass 3.3.
- Interchangeable valve spindle
- Spindle with retention device
- Lateral or straight needle valve available
Valve housing with precision burette tip, PTFE spindle with PP cap.
Frequently Bought Together