Stopcocks for burettes, lateral-valve
Catalog # HIRS8901450
Supplier: HIRSCHMANN
Specifications
- For:50 ml burettes
- Seq:03
- Pk:1
Specifications
About this item
Lateral burette stopcock with PTFE spindle, made of DURAN®, borosilicate glass 3.3, for burettes, clear glass.
- Excellent chemical resistance
- Excellent temperature resistance